Assay Details
| Gene Symbol: TEX36-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_023362.1 | 4 - 5 | 506 | 91 | ||
| GenBank mRNA | BC033977.2 | - | 4 - 5 | 506 | 91 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000910.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16394173 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27942329 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27942299 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07551477 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27942321 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07551496 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16394176 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27942295 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27942296 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27942298 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07551476 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27942320 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07551495 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27942297 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07551485 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07551465 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07551466 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07551484 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07551468 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07551482 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27942291 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16394170 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27942315 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27942305 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27942312 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27942322 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27942306 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16394172 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07551487 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07551499 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07551492 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07551493 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27942331 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 72556 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 560734 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 192059 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P120127 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P120127 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 35380 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 35036 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P02365869 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P120127 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_32_P120127 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16394173 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07551477 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07551485 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27942300 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27942305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07551482 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27942315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07551476 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07551484 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16394172 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27942312 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27942297 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07551487 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07551481 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16394170 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07551468 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27942295 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27942299 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07551478 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16394170 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07551477 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16394173 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07551482 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27942305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27942295 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07551487 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16394172 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07551468 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07551476 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27942312 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07551484 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27942300 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07551481 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07551478 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27942299 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27942297 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07551485 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27942315 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P120127 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P02365869 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P120127 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P39216302 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02365869 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P120131 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551476 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551514 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16394175 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551537 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551487 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551466 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551504 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551516 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551508 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551525 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551534 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551495 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551528 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551489 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551490 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551535 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551474 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551468 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551481 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551482 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551492 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551484 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551531 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551511 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551517 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551472 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551477 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551498 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551499 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551502 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16394171 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551479 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551532 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551523 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551529 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551485 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551520 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551506 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07551465 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02365869 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1556133_PM_s_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000431861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000431861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000431861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000431861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009237.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011137.hg.1 | ENST00000431861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011137.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011137.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011137.hg.1 | TC10000910.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011138.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011138.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011138.hg.1 | TC10000910.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000163305.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163305.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163305.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000163305.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163305.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163305.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000163305.hg.1 | TC1000009237.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000431861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000431861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000431861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000431861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009237.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000163304.hg.1 | ENST00000431861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163304.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163304.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163304.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000163304.hg.1 | ENST00000431861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163304.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163304.hg.1 | ENST00000446081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163304.hg.1 | NR_023362.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000163304.hg.1 | TC1000009237.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1556133_PM_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1556133_s_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556133_s_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556133_s_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556133_s_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556133_s_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556133_s_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556133_s_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556133_s_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556133_s_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556133_s_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556133_s_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556133_s_at1 | - | - |
| Human Genome U95 Set | 74465_at | - | - |
| Human Genome U95 Set | 74465_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044625 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044625 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 199334 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3247792 | NR_023362.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3247792 | BC033977.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3247792 | NR_023362.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3247792 | BC033977.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3247792 | NR_023362.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3247792 | BC033977.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3247792 | NR_023362.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3247792 | BC033977.2 | human_transcriptome |

TEX36-AS1
Genomic Map