Assay Details
| Gene Symbol: ADCY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021116.2 | NP_066939.1 | 10 - 11 | 1911 | 81 | |
| XM_005249584.3 | XP_005249641.1 | 10 - 11 | 1909 | 81 | ||
| GenBank mRNA | AB209496.1 | - | 10 - 11 | 1856 | 81 | |
| L05500.1 | - | 8 - 9 | 1055 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000283.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
renal water homeostasis
cAMP biosynthetic process
adenylate cyclase-activating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
axonogenesis
long-term memory
circadian rhythm
response to lithium ion
cAMP-mediated signaling
activation of protein kinase A activity
response to drug
regulation of circadian rhythm
cellular response to calcium ion
cellular response to glucagon stimulus
cellular response to forskolin
adenylate cyclase activity
calmodulin binding
ATP binding
calcium- and calmodulin-responsive adenylate cyclase activity
metal ion binding
renal water homeostasis
cAMP biosynthetic process
adenylate cyclase-activating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
axonogenesis
long-term memory
circadian rhythm
response to lithium ion
cAMP-mediated signaling
activation of protein kinase A activity
response to drug
regulation of circadian rhythm
cellular response to calcium ion
cellular response to glucagon stimulus
cellular response to forskolin
adenylate cyclase activity
calmodulin binding
ATP binding
calcium- and calmodulin-responsive adenylate cyclase activity
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07004503.hg.1 | TC07000283.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004503.hg.1 | ENST00000297323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004503.hg.1 | ENST00000545300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004503.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000468353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000468353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000468353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000468353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000468353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000468353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700151688.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700151688.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700151688.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700151688.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700151688.hg.1 | TC0700007426.hg.1 | Clariom D TC |

ADCY1
Genomic Map