Assay Details
| Gene Symbol: LAMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005559.3 | NP_005550.2 | 53 - 54 | 7720 | 106 | |
| XM_011525655.2 | XP_011523957.1 | 53 - 54 | 7714 | 106 | ||
| XM_011525656.2 | XP_011523958.1 | 43 - 44 | 6135 | 106 | ||
| GenBank mRNA | AK091949.1 | - | 4 - 5 | 1033 | 106 | |
| BC039051.1 | - | 5 - 6 | 644 | 106 | ||
| BC062318.1 | - | 1 - 2 | 80 | 106 | ||
| BC144590.1 | - | 52 - 53 | 8641 | 106 | ||
| X58531.1 | - | 53 - 54 | 7720 | 106 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000309.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK091949.1 transcripts.
Set Membership: |
|

LAMA1
Genomic Map