Assay Details
| Gene Symbol: GCC2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135290.1 | 2 - 3 | 476 | 78 | ||
| GenBank mRNA | BC004487.2 | - | 2 - 3 | 453 | 3832541 | 78 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21499705 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21499706 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15298639 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21499708 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21499703 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3325871 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P169892 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P16806 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 9750 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 23349 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 18039 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3325871 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P169892 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P16806 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P16806 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P169892 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 29795 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 62174 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P132940 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15776448 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00441818 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | XR_040183_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | XR_040182_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | XR_040184_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8054450 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8054449 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 224141_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013806.hg.1 | ENST00000322353.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013806.hg.1 | ENST00000322353.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013806.hg.1 | ENST00000322353.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013806.hg.1 | ENST00000322353.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013806.hg.1 | ENST00000440975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013806.hg.1 | ENST00000440975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013806.hg.1 | ENST00000440975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201131.hg.1 | ENST00000322353.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201131.hg.1 | ENST00000322353.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201131.hg.1 | TC0200013806.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200013806.hg.1 | ENST00000322353.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013806.hg.1 | ENST00000322353.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013806.hg.1 | ENST00000322353.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013806.hg.1 | ENST00000322353.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013806.hg.1 | ENST00000440975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013806.hg.1 | ENST00000440975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013806.hg.1 | ENST00000440975.1 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 224141_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 224141_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224141_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224141_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224141_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224141_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224141_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224141_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224141_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224141_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224141_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224141_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224141_at11 | - | - |
| Human Genome U133 Set | 224141_PM_at | - | - |
| Human Genome U133 Set | 224141_at | - | - |

GCC2-AS1
Genomic Map