Assay Details
| Gene Symbol: FAM49A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030797.3 | NP_110424.1 | 7 - 8 | 771 | 74 | |
| XM_005262630.3 | XP_005262687.1 | 6 - 7 | 727 | 74 | ||
| XM_006712108.3 | XP_006712171.1 | 7 - 8 | 726 | 74 | ||
| GenBank mRNA | AK055334.1 | - | 7 - 8 | 739 | 74 | |
| AK091280.1 | - | 2 - 3 | 185 | 74 | ||
| AK315065.1 | - | 6 - 7 | 666 | 74 | ||
| AL136704.1 | - | 7 - 8 | 736 | 74 | ||
| BC038971.1 | - | 7 - 8 | 722 | 5200807 | 74 | |
| CR533504.1 | - | 5 - 6 | 518 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001600.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02025672.hg.1 | TC02001600.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025672.hg.1 | ENST00000355549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025672.hg.1 | ENST00000381323.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025672.hg.1 | ENST00000406434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025672.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025673.hg.1 | TC02001600.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025673.hg.1 | ENST00000355549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025673.hg.1 | ENST00000381323.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025673.hg.1 | ENST00000406434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025673.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025674.hg.1 | TC02001600.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025674.hg.1 | ENST00000355549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025674.hg.1 | ENST00000381323.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025674.hg.1 | ENST00000406434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025674.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000445605.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000445605.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000445605.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000445605.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000445605.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000451689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000451689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000451689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000451689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000451689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000451689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000469507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000469507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000469507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | ENST00000469507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200185943.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200185943.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200185943.hg.1 | ENST00000469507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200185943.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200185943.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200185943.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200185943.hg.1 | ENST00000469507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200185943.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200185943.hg.1 | TC0200011837.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200185944.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200185944.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200185944.hg.1 | ENST00000469507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200185944.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200185944.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200185944.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200185944.hg.1 | ENST00000469507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200185944.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200185944.hg.1 | TC0200011837.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000445605.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000445605.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000445605.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000445605.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000445605.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000451689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000451689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000451689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000451689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000451689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000451689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000469507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000469507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000469507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | ENST00000469507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011837.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200185945.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200185945.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200185945.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200185945.hg.1 | ENST00000381323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200185945.hg.1 | ENST00000406434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200185945.hg.1 | NM_030797.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200185945.hg.1 | TC0200011837.hg.1 | Clariom D TC |

FAM49A
Genomic Map