| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009429.hg.1 |
ENST00000484765.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009429.hg.1 |
ENST00000484765.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009429.hg.1 |
ENST00000484765.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009429.hg.1 |
ENST00000484765.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009429.hg.1 |
ENST00000484765.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009429.hg.1 |
NR_109989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009429.hg.1 |
NR_109989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009429.hg.1 |
NR_109989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009429.hg.1 |
NR_109989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009429.hg.1 |
NR_109989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009429.hg.1 |
NR_109989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009429.hg.1 |
ENST00000484765.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009429.hg.1 |
ENST00000484765.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009429.hg.1 |
ENST00000484765.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009429.hg.1 |
ENST00000484765.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009429.hg.1 |
ENST00000484765.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009429.hg.1 |
NR_109989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009429.hg.1 |
NR_109989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009429.hg.1 |
NR_109989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009429.hg.1 |
NR_109989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009429.hg.1 |
NR_109989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009429.hg.1 |
NR_109989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300167773.hg.1 |
ENST00000484765.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300167773.hg.1 |
NR_109989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300167773.hg.1 |
ENST00000484765.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300167773.hg.1 |
NR_109989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300167773.hg.1 |
TC0300009429.hg.1 |
Clariom D TC |