Assay Details
| Gene Symbol: TMEM179B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199337.2 | NP_955369.1 | 1 - 2 | 129 | 85 | |
| XM_005273982.3 | XP_005274039.3 | 1 - 2 | 210 | 85 | ||
| GenBank mRNA | BC051355.1 | - | 1 - 2 | 104 | 6597282 | 85 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000565.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11006817.hg.1 | ENST00000333449.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006817.hg.1 | ENST00000533861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006817.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006817.hg.1 | TC11000565.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006816.hg.1 | ENST00000333449.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006816.hg.1 | ENST00000533861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006816.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006816.hg.1 | TC11000565.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000526546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000526546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000526546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000532345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000532345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000532345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000532345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000532345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000532345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000532345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000532586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000532586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000532586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000532586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000532586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000532586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000532586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000532586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | ENST00000610433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007876.hg.1 | NR_106806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000526546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000526546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000526546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000532345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000532345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000532345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000532345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000532345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000532345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000532345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000532586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000532586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000532586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000532586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000532586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000532586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000532586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000532586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | ENST00000610433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007876.hg.1 | NR_106806.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100156118.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156118.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156118.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100156118.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156118.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156118.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100156118.hg.1 | TC1100007876.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100156119.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156119.hg.1 | ENST00000532345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156119.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156119.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100156119.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156119.hg.1 | ENST00000532345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156119.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156119.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100156119.hg.1 | TC1100007876.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100156117.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156117.hg.1 | ENST00000532586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156117.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156117.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100156117.hg.1 | ENST00000333449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156117.hg.1 | ENST00000532586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156117.hg.1 | ENST00000533861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156117.hg.1 | NM_199337.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100156117.hg.1 | TC1100007876.hg.1 | Clariom D TC |

TMEM179B
Genomic Map