Assay Details
| Gene Symbol: BCAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024050.1 | 1 - 2 | 130 | 80 | ||
| GenBank mRNA | BX107175.1 | - | 1 - 2 | 101 | 80 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000873.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000571259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000573037.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000573037.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000573037.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000573037.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000573037.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000573037.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000573037.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000573319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000573319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000573319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000573319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000573319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000573319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000573319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000573319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000573319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000573319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000574028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000574028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000574028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000577041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000577041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000577041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000615574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000615574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | ENST00000615574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_024049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_024049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_024049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_024049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_024049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_024049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_024049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_024049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_024049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_024049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_024050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_024050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_024050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_024050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_024050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_024050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_131216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_131216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_131216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_131216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_131216.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_131217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_131217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_131217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_131217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_131217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_131217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_131217.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_131222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_131222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_131222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_131222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_131222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009419.hg.1 | NR_131222.1 | Refseq |

BCAR4
Genomic Map