Assay Details
| Gene Symbol: LINC01057 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104131.1 | 2 - 3 | 506 | 86 | ||
| GenBank mRNA | BC030750.1 | - | 2 - 3 | 512 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01057| Gene Symbol: LINC01057 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104131.1 | 2 - 3 | 506 | 86 | ||
| GenBank mRNA | BC030750.1 | - | 2 - 3 | 512 | 86 | |
Set Membership: |
|