Assay Details
| Gene Symbol: DNLZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080849.2 | NP_001074318.1 | 2 - 3 | 80 | ||
| NR_073565.1 | 2 - 3 | 80 | ||||
| GenBank mRNA | AI435052.1 | - | 1 - 2 | 278 | 80 | |
| BC146915.1 | - | 2 - 3 | 9056843 | 80 | ||
| BC146924.1 | - | 2 - 3 | 9056852 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09002925.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07008172 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27358914 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07008174 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07008171 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07008167 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07008173 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07008178 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07008168 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07008163 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07008176 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07008169 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07008165 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27358924 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 194599 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 298990 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 205618 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3358403 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3243264 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P216836 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 45039 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 38611 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 19342 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P02194043 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P32578431 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3243264 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3358403 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3243264 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3358403 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07008173 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07008178 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07008172 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07008171 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27358924 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07008169 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07008176 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07008174 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07008178 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07008173 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07008169 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07008176 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27358924 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07008174 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07008172 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07008171 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P216836 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P32578431 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008232 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008134 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008174 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16276802 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16276719 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008126 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16276803 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008172 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008171 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008217 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008219 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16276757 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008139 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008168 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008234 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008230 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008166 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008119 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008138 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16276723 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16276742 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008215 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008122 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008228 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008128 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008133 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008147 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008223 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008120 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008220 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16276752 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008178 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16276710 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008237 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008135 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008132 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008214 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008141 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008125 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16276755 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16276799 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008222 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008213 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008167 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008146 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008169 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008236 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16276720 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008129 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16276795 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16276759 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16276762 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16276728 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008144 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008225 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16276702 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07008176 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001080849_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8165111 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8165108 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8165112 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8165109 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8165107 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 228272_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11728462_at | - | - |
| Affymetrix Human Genome U219 Array | 11728461_PM_a_at | - | - |
| Affymetrix Human Genome U219 Array | 11728461_a_at | - | - |
| Affymetrix Human Genome U219 Array | 11728462_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022072.hg.1 | TC09002925.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022072.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022072.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022066.hg.1 | TC09002925.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022066.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022066.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022066.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022067.hg.1 | TC09002925.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022067.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022067.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022067.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022071.hg.1 | TC09002925.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022071.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022071.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022071.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022068.hg.1 | TC09002925.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022068.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022068.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186816.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186816.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186816.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186816.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186816.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186816.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186816.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186816.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186816.hg.1 | TC0900012284.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900186817.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186817.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186817.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186817.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186817.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186817.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186817.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186817.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186817.hg.1 | TC0900012284.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900186818.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186818.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186818.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186818.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186818.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186818.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186818.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186818.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186818.hg.1 | TC0900012284.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 228272_at | - | - |
| Human Genome U133 Plus 2.0 Array | 228272_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228272_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228272_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228272_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228272_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228272_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228272_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228272_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228272_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228272_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228272_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228272_at11 | - | - |
| Human Genome U95 Set | 90264_at | - | - |
| Human Genome U95 Set | 90264_PM_at | - | - |
| Human Genome U133 Set | 228272_PM_at | - | - |
| Human Genome U133 Set | 228272_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A038214 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A038214 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2112049 | NM_001080849.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2112049 | NM_001080849.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2112049 | NM_001080849.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2112049 | NM_001080849.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2112049 | NM_001080849.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2112049 | NM_001080849.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2112049 | NM_001080849.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2112049 | NM_001080849.1 | human_transcriptome |

DNLZ
Genomic Map