Assay Details
| Gene Symbol: TRIM59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173084.2 | NP_775107.1 | 1 - 2 | 124 | 91 | |
| GenBank mRNA | AK291284.1 | - | 1 - 2 | 46 | 91 | |
| AY159379.1 | - | 1 - 2 | 124 | 91 | ||
| BC109259.1 | - | 1 - 2 | 111 | 40025031 | 91 | |
| BC109260.1 | - | 1 - 2 | 111 | 40025036 | 91 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03003399.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK291284.1,AY159379.1,BC109259.1,BC109260.1,NM_173084.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03034982.hg.1 | TC03003399.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03034982.hg.1 | ENST00000309784.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03034982.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03034975.hg.1 | TC03003399.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03034975.hg.1 | ENST00000309784.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03034975.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03034975.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000468542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000468542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000471155.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000471155.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000471155.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000471155.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000471155.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000471396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000471396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000471396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000471396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000471396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000479460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000479460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000479460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000479460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000479460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000479460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000479460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000479460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000494486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000494486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000494486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000494486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000494486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000496222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000496222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000496222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000496222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000496222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000496222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03034985.hg.1 | TC03003399.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03034985.hg.1 | ENST00000309784.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03034985.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300209517.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209517.hg.1 | ENST00000471155.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209517.hg.1 | ENST00000479460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209517.hg.1 | ENST00000494486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209517.hg.1 | ENST00000496222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209517.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209517.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300209517.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209517.hg.1 | ENST00000471155.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209517.hg.1 | ENST00000479460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209517.hg.1 | ENST00000494486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209517.hg.1 | ENST00000496222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209517.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209517.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300209517.hg.1 | TC0300014075.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000468542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000468542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000471155.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000471155.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000471155.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000471155.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000471155.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000471396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000471396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000471396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000471396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000471396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000479460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000479460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000479460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000479460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000479460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000479460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000479460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000479460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000494486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000494486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000494486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000494486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000494486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000496222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000496222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000496222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000496222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000496222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000496222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | ENST00000543469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014075.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300209524.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209524.hg.1 | ENST00000471155.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209524.hg.1 | ENST00000471396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209524.hg.1 | ENST00000496222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209524.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300209524.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209524.hg.1 | ENST00000471155.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209524.hg.1 | ENST00000471396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209524.hg.1 | ENST00000496222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209524.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300209524.hg.1 | TC0300014075.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300209527.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209527.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300209527.hg.1 | ENST00000309784.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209527.hg.1 | NM_173084.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300209527.hg.1 | TC0300014075.hg.1 | Clariom D TC |

TRIM59
Genomic Map