Assay Details
| Gene Symbol: LOC102723362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110577.1 | 2 - 3 | 228 | 75 | ||
| GenBank mRNA | AA781119.1 | - | 1 - 2 | 75 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723362| Gene Symbol: LOC102723362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110577.1 | 2 - 3 | 228 | 75 | ||
| GenBank mRNA | AA781119.1 | - | 1 - 2 | 75 | ||
Set Membership: |
|