Assay Details
| Gene Symbol: VMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182566.2 | NP_872372.1 | 2 - 3 | 394 | 64 | |
| GenBank mRNA | AF521892.1 | - | 2 - 3 | 399 | 64 | |
| AY359099.1 | - | 2 - 3 | 324 | 64 | ||
| BC104194.2 | - | 2 - 3 | 189 | 40031204 | 64 | |
| BC104195.1 | - | 2 - 3 | 362 | 40031205 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000328739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000328739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000328739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000328739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000328739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000328739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000328739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000354194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000354194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000354194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000354194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000354194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000354194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000416307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000416307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000416307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000416307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000416307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000416307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000441199.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000441199.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000441199.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000441199.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000441199.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | ENST00000441199.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144940.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144940.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144940.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144940.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144940.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144940.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144940.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_001144941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_182566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_182566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_182566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_182566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_182566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_182566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009530.hg.1 | NM_182566.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173339.hg.1 | ENST00000328739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173339.hg.1 | ENST00000354194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173339.hg.1 | ENST00000416307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173339.hg.1 | ENST00000441199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173339.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173339.hg.1 | NM_001144940.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173339.hg.1 | NM_001144941.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173339.hg.1 | NM_182566.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173339.hg.1 | ENST00000328739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173339.hg.1 | ENST00000354194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173339.hg.1 | ENST00000416307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173339.hg.1 | ENST00000441199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173339.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173339.hg.1 | NM_001144940.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173339.hg.1 | NM_001144941.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173339.hg.1 | NM_182566.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173339.hg.1 | TC1700009530.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700173340.hg.1 | ENST00000328739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173340.hg.1 | ENST00000354194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173340.hg.1 | ENST00000416307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173340.hg.1 | ENST00000441199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173340.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173340.hg.1 | NM_001144940.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173340.hg.1 | NM_001144941.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173340.hg.1 | NM_182566.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173340.hg.1 | ENST00000328739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173340.hg.1 | ENST00000354194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173340.hg.1 | ENST00000416307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173340.hg.1 | ENST00000441199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173340.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173340.hg.1 | NM_001144940.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173340.hg.1 | NM_001144941.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173340.hg.1 | NM_182566.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173340.hg.1 | TC1700009530.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000328739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000328739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000328739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000328739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000328739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000328739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000328739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000354194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000354194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000354194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000354194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000354194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000354194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000416307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000416307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000416307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000416307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000416307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000416307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000441199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000441199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000441199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000441199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000441199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | ENST00000441199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144940.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144940.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144940.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144940.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144940.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144940.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144940.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_001144941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_182566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_182566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_182566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_182566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_182566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_182566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009530.hg.1 | NM_182566.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 201029 | - | - |

VMO1
Genomic Map