Assay Details
| Gene Symbol: HBBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_001589.1 | 2 - 3 | 361 | 88 | ||
| GenBank mRNA | N55205.1 | - | 1 - 2 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001332.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P732099 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P732099 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 33642 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 7375 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P732099 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P732099 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_19_P00813593 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012660 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012657 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613537 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613553 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16415862 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012662 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012651 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012663 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613572 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613570 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613574 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012666 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613555 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012642 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613547 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613538 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012676 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613548 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613563 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613575 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012640 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012640 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613553 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613563 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012660 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613572 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012657 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613547 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012666 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613570 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613555 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012651 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613574 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012662 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613548 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613537 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012663 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012642 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613538 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16415862 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07613575 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28012676 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P732099 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P732099 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613564 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613478 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613494 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613480 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613577 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613474 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613550 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613542 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613593 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613508 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16415886 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613568 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613588 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613476 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16415889 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613499 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613552 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613503 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613572 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613591 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613544 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613538 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613575 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613539 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613584 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613535 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16415887 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613506 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613556 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613500 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613548 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613559 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613537 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613562 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613501 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613583 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613482 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613570 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613545 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613554 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613486 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613487 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613492 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613566 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07613484 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 216063_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR11016760.hg.1 | ENST00000454892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11016760.hg.1 | NR_001589.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11016760.hg.1 | TC11001332.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009919.hg.1 | ENST00000433329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009919.hg.1 | ENST00000433329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009919.hg.1 | ENST00000433329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009919.hg.1 | ENST00000433329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009919.hg.1 | ENST00000454892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009919.hg.1 | ENST00000454892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009919.hg.1 | ENST00000454892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009919.hg.1 | NR_001589.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009919.hg.1 | NR_001589.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009919.hg.1 | NR_001589.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009919.hg.1 | NR_001589.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009919.hg.1 | NR_001589.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009919.hg.1 | NR_001589.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100174868.hg.1 | ENST00000454892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100174868.hg.1 | NR_001589.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100174868.hg.1 | ENST00000454892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100174868.hg.1 | NR_001589.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100174868.hg.1 | TC1100009919.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100174869.hg.1 | ENST00000433329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100174869.hg.1 | ENST00000454892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100174869.hg.1 | NR_001589.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100174869.hg.1 | ENST00000433329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100174869.hg.1 | ENST00000454892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100174869.hg.1 | NR_001589.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100174869.hg.1 | TC1100009919.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100009919.hg.1 | ENST00000433329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009919.hg.1 | ENST00000433329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009919.hg.1 | ENST00000433329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009919.hg.1 | ENST00000433329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009919.hg.1 | ENST00000454892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009919.hg.1 | ENST00000454892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009919.hg.1 | ENST00000454892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009919.hg.1 | NR_001589.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009919.hg.1 | NR_001589.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009919.hg.1 | NR_001589.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009919.hg.1 | NR_001589.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009919.hg.1 | NR_001589.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009919.hg.1 | NR_001589.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 216063_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216063_at | - | - |
| Human Genome U133A 2.0 Array | 216063_PM_at | - | - |
| Human Genome U133A 2.0 Array | 216063_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NR_001589_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw583526 | - | - |
| Human Genome U95 Set | 35920_PM_at | - | - |
| Human Genome U95 Set | 35920_at | - | - |
| Human Genome U95 Set | 35921_at | - | - |
| Human Genome U95 Set | 35921_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216063_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216063_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216063_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216063_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216063_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216063_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216063_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216063_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216063_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216063_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216063_at11 | - | - |
| Human Genome U133 Set | 216063_PM_at | - | - |
| Human Genome U133 Set | 216063_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044836 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044837 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044836 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044837 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 124304 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1653814 | NR_001589.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1653814 | N55205.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1653814 | NR_001589.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1653814 | N55205.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1653814 | NR_001589.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1653814 | N55205.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1653814 | NR_001589.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1653814 | N55205.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1653814 | NR_001589.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1653814 | N55205.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1653814 | NR_001589.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1653814 | N55205.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 7100747 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_12345 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1653814 | NR_001589.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1653814 | N55205.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1653814 | NR_001589.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1653814 | N55205.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1653814 | NR_001589.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1653814 | N55205.1 | human_transcriptome |

HBBP1
Genomic Map