Assay Details
| Gene Symbol: LOC100289673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033175.1 | 2 - 3 | 201 | 86 | ||
| GenBank mRNA | AI125295.1 | - | 1 - 2 | 86 | ||
| BX093587.1 | - | 2 - 3 | 200 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000516.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364311 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364312 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364326 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184804 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364344 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364333 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15730218 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364280 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364321 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364309 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364310 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364278 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184852 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364334 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15730221 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364319 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184876 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364318 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364346 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15730223 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364343 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184813 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364341 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364308 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364297 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364295 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364324 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364320 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364353 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364338 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364348 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364284 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184798 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364352 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184803 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184823 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184863 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364327 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364290 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364298 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184811 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364344 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364297 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364343 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364341 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184876 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184803 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364311 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364312 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184804 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15730218 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15730221 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364308 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364284 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364318 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184811 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15730223 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184798 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364346 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184863 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364321 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364295 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364320 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364309 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184852 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364353 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184823 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364278 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364352 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364333 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364327 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364324 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364338 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364298 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184813 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364310 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04184810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364334 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364326 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364280 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364348 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364319 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364290 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24364315 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05007161.hg.1 | TC05000516.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05007161.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011681.hg.1 | ENST00000518923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05007159.hg.1 | TC05000516.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05007159.hg.1 | ENST00000523446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05007159.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008298.hg.1 | ENST00000523446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008298.hg.1 | ENST00000523446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008298.hg.1 | ENST00000523446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008298.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008298.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008298.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008298.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05007160.hg.1 | TC05000516.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05007160.hg.1 | ENST00000523446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05007160.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05007158.hg.1 | TC05000516.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05007158.hg.1 | ENST00000523446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05007158.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500156753.hg.1 | ENST00000523446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500156753.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500156753.hg.1 | ENST00000523446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500156753.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500156753.hg.1 | TC0500008298.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500008298.hg.1 | ENST00000523446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008298.hg.1 | ENST00000523446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008298.hg.1 | ENST00000523446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008298.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008298.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008298.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008298.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500156754.hg.1 | ENST00000523446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500156754.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500156754.hg.1 | ENST00000523446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500156754.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500156754.hg.1 | TC0500008298.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500011681.hg.1 | ENST00000518923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500156756.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500156756.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500156756.hg.1 | TC0500008298.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500156755.hg.1 | ENST00000523446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500156755.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500156755.hg.1 | ENST00000523446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500156755.hg.1 | NR_033175.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500156755.hg.1 | TC0500008298.hg.1 | Clariom D TC |
| Human Genome U95 Set | 51417_at | - | - |
| Human Genome U95 Set | 51417_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A028604 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A028604 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 165039 | - | - |

LOC100289673
Genomic Map