| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
ENST00000464242.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
ENST00000464242.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
ENST00000464242.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
ENST00000464242.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
ENST00000468377.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
ENST00000468377.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
ENST00000468377.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
ENST00000468377.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
ENST00000468377.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
ENST00000468377.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
ENST00000473958.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
ENST00000473958.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
ENST00000473958.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
NR_125398.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
NR_125398.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
NR_125398.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
NR_125398.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
NR_125398.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008724.hg.1 |
NR_125398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162040.hg.1 |
ENST00000468377.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162040.hg.1 |
NR_125398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162040.hg.1 |
ENST00000468377.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162040.hg.1 |
NR_125398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162040.hg.1 |
TC0300008724.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
ENST00000464242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
ENST00000464242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
ENST00000464242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
ENST00000464242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
ENST00000468377.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
ENST00000468377.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
ENST00000468377.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
ENST00000468377.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
ENST00000468377.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
ENST00000468377.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
ENST00000473958.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
ENST00000473958.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
ENST00000473958.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
NR_125398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
NR_125398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
NR_125398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
NR_125398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
NR_125398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008724.hg.1 |
NR_125398.1 |
Refseq |