Assay Details
| Gene Symbol: LINC01111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105006.1 | 2 - 3 | 229 | 144 | ||
| GenBank mRNA | BC015449.1 | - | 2 - 3 | 229 | 144 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01111| Gene Symbol: LINC01111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105006.1 | 2 - 3 | 229 | 144 | ||
| GenBank mRNA | BC015449.1 | - | 2 - 3 | 229 | 144 | |
Set Membership: |
|