Assay Details
| Gene Symbol: LINC01012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038293.1 | 1 - 2 | 466 | 73 | ||
| NR_038294.1 | 1 - 2 | 466 | 73 | |||
| GenBank mRNA | AL120625.1 | - | 1 - 2 | 467 | 73 | |
| BI771077.1 | - | 1 - 2 | 453 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000260.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629578 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15796381 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819691 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819689 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629627 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629568 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819718 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629649 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819711 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629582 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629643 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629577 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629620 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629589 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15796380 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629650 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629588 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15796388 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819665 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629613 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629628 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819690 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819714 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15796387 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629585 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629630 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819743 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629584 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629607 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629646 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819710 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819724 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629611 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629641 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629580 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629642 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629599 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629608 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819685 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629597 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819712 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629645 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819695 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819671 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819667 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819747 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819700 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629633 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819686 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629598 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629634 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629652 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629579 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819696 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629576 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629647 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819694 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819716 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629571 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819735 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819698 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629567 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629612 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819734 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04629615 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24819717 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P37558465 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06002200.hg.1 | TC06000260.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002200.hg.1 | NR_038293.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002200.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000625477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000625477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000625477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000625477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000628328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000628328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000628328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000628328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000628762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000628762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000628762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000628762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000628762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | NR_038293.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | NR_038293.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | NR_038293.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | NR_038293.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007367.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002203.hg.1 | TC06000260.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002203.hg.1 | ENST00000446316.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002203.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002203.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002206.hg.1 | TC06000260.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002206.hg.1 | NR_038293.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002206.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002198.hg.1 | TC06000260.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002198.hg.1 | NR_038293.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002198.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002201.hg.1 | TC06000260.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002201.hg.1 | ENST00000446316.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002201.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002201.hg.1 | NR_038293.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002201.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149943.hg.1 | ENST00000625477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149943.hg.1 | ENST00000628328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149943.hg.1 | ENST00000628762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149943.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149943.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149943.hg.1 | NR_038293.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149943.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149943.hg.1 | ENST00000625477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149943.hg.1 | ENST00000628328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149943.hg.1 | ENST00000628762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149943.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149943.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149943.hg.1 | NR_038293.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149943.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149943.hg.1 | TC0600007367.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600149941.hg.1 | ENST00000625477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149941.hg.1 | ENST00000628328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149941.hg.1 | ENST00000628762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149941.hg.1 | NR_038293.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149941.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149941.hg.1 | ENST00000625477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149941.hg.1 | ENST00000628328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149941.hg.1 | ENST00000628762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149941.hg.1 | NR_038293.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149941.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149941.hg.1 | TC0600007367.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600149954.hg.1 | ENST00000628328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149954.hg.1 | ENST00000628762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149954.hg.1 | NR_038293.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149954.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149954.hg.1 | ENST00000628328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149954.hg.1 | ENST00000628762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149954.hg.1 | NR_038293.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149954.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149954.hg.1 | TC0600007367.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000625477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000625477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000625477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000625477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000628328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000628328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000628328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000628328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000628762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000628762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000628762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000628762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000628762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | NR_038293.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | NR_038293.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | NR_038293.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | NR_038293.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007367.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149950.hg.1 | ENST00000628762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149950.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149950.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149950.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149950.hg.1 | ENST00000628762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149950.hg.1 | ENST00000629732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149950.hg.1 | NR_038292.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149950.hg.1 | NR_038294.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149950.hg.1 | TC0600007367.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1568826_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1568826_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1568826_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1568826_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1568826_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1568826_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1568826_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1568826_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1568826_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1568826_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1568826_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1568826_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1568826_at11 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031149 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031149 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 126755 | - | - |

LINC01012
Genomic Map