| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006952.hg.1 |
ENST00000542689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006952.hg.1 |
ENST00000542689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006952.hg.1 |
ENST00000542689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006952.hg.1 |
ENST00000542689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006952.hg.1 |
NR_120466.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006952.hg.1 |
NR_120466.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006952.hg.1 |
NR_120466.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006952.hg.1 |
NR_120466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200148592.hg.1 |
ENST00000542689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200148592.hg.1 |
NR_120466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200148592.hg.1 |
ENST00000542689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200148592.hg.1 |
NR_120466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200148592.hg.1 |
TC1200006952.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200148591.hg.1 |
ENST00000542689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200148591.hg.1 |
NR_120466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200148591.hg.1 |
ENST00000542689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200148591.hg.1 |
NR_120466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200148591.hg.1 |
TC1200006952.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200148593.hg.1 |
ENST00000542689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200148593.hg.1 |
NR_120466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200148593.hg.1 |
ENST00000542689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200148593.hg.1 |
NR_120466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200148593.hg.1 |
TC1200006952.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200006952.hg.1 |
ENST00000542689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006952.hg.1 |
ENST00000542689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006952.hg.1 |
ENST00000542689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006952.hg.1 |
ENST00000542689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006952.hg.1 |
NR_120466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006952.hg.1 |
NR_120466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006952.hg.1 |
NR_120466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006952.hg.1 |
NR_120466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200148590.hg.1 |
ENST00000542689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200148590.hg.1 |
NR_120466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200148590.hg.1 |
ENST00000542689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200148590.hg.1 |
NR_120466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200148590.hg.1 |
TC1200006952.hg.1 |
Clariom D TC |