Assay Details
| Gene Symbol: ANTXRLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103828.1 | 2 - 3 | 296 | 74 | ||
| GenBank mRNA | BX091290.1 | - | 2 - 3 | 341 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANTXRLP1| Gene Symbol: ANTXRLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103828.1 | 2 - 3 | 296 | 74 | ||
| GenBank mRNA | BX091290.1 | - | 2 - 3 | 341 | 74 | |
Set Membership: |
|