Assay Details
Gene Symbol: LOC200726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102659.1 | NP_001096129.1 | 2 - 3 | 583 | 144 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001220.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02018723.hg.1 | TC02001220.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02018723.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02018723.hg.1 | ENST00000453816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02018723.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02018723.hg.1 | NM_001102659.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02018724.hg.1 | TC02001220.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02018724.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02018724.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02018724.hg.1 | NM_001102659.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | ENST00000453816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | ENST00000453816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | NM_001102659.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | NM_001102659.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | NM_001102659.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | NM_001102659.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010580.hg.1 | NM_001102659.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | ENST00000453816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | ENST00000453816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | NM_001102659.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | NM_001102659.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | NM_001102659.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | NM_001102659.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010580.hg.1 | NM_001102659.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200174940.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174940.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174940.hg.1 | ENST00000453816.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174940.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174940.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174940.hg.1 | NM_001102659.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200174940.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174940.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174940.hg.1 | ENST00000453816.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174940.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174940.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174940.hg.1 | NM_001102659.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200174940.hg.1 | TC0200010580.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200174941.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174941.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174941.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174941.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174941.hg.1 | NM_001102659.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200174941.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174941.hg.1 | ENST00000441223.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174941.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174941.hg.1 | ENST00000543490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200174941.hg.1 | NM_001102659.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200174941.hg.1 | TC0200010580.hg.1 | Clariom D TC |
