Assay Details
| Gene Symbol: NPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000906.3 | NP_000897.3 | 10 - 11 | 2179 | 80 | |
| XM_017001374.1 | XP_016856863.1 | 10 - 11 | 2250 | 80 | ||
| GenBank mRNA | AK025024.1 | - | 10 - 11 | 2179 | 80 | |
| AK300446.1 | - | 9 - 10 | 1385 | 80 | ||
| BC063304.1 | - | 10 - 11 | 2228 | 30331757 | 80 | |
| X15357.1 | - | 10 - 11 | 1801 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001262.hg.1
Set Membership: |
|

NPR1
Genomic Map