| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007604.hg.1 |
ENST00000454388.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007604.hg.1 |
ENST00000454388.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007604.hg.1 |
ENST00000454388.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007604.hg.1 |
ENST00000454388.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010049.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010049.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010049.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010049.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010049.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010049.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010049.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010049.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010049.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010049.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168619.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168619.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168619.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168619.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168619.hg.1 |
TC0X00010049.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00168617.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168617.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168617.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168617.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168617.hg.1 |
TC0X00010049.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00168615.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168615.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168615.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168615.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168615.hg.1 |
TC0X00010049.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00168618.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168618.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168618.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168618.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168618.hg.1 |
TC0X00010049.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00010049.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010049.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010049.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010049.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010049.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010049.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010049.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010049.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010049.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010049.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168616.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168616.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168616.hg.1 |
ENST00000416989.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168616.hg.1 |
NR_110398.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168616.hg.1 |
TC0X00010049.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00007604.hg.1 |
ENST00000454388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007604.hg.1 |
ENST00000454388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007604.hg.1 |
ENST00000454388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007604.hg.1 |
ENST00000454388.1 |
Ensembl |