Assay Details
| Gene Symbol: LOC101929570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110258.2 | 2 - 3 | 113 | 94 | ||
| GenBank mRNA | BC042876.1 | - | 2 - 3 | 113 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929570| Gene Symbol: LOC101929570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110258.2 | 2 - 3 | 113 | 94 | ||
| GenBank mRNA | BC042876.1 | - | 2 - 3 | 113 | 94 | |
Set Membership: |
|