Assay Details
| Gene Symbol: RAB15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308154.1 | NP_001295083.1 | 2 - 3 | 266 | 80 | |
| NM_198686.2 | NP_941959.1 | 2 - 3 | 266 | 80 | ||
| XM_017021277.1 | XP_016876766.1 | 3 - 4 | 526 | 80 | ||
| GenBank mRNA | AB672631.2 | - | 2 - 3 | 235 | 80 | |
| AB672632.1 | - | 2 - 3 | 236 | 80 | ||
| AB678452.1 | - | 2 - 3 | 266 | 80 | ||
| AB678453.1 | - | 2 - 3 | 249 | 80 | ||
| BC040679.2 | - | 2 - 3 | 189 | 4817835 | 80 | |
| BX248046.1 | - | 1 - 2 | 62 | 80 | ||
| BX640825.1 | - | 2 - 3 | 149 | 80 | ||
| CN352234.1 | - | 2 - 3 | 267 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001224.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14013953.hg.1 | ENST00000267512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14013953.hg.1 | ENST00000426039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14013953.hg.1 | ENST00000436278.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14013953.hg.1 | ENST00000533601.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14013953.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14013953.hg.1 | TC14001224.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400169645.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400169645.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400169645.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400169645.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400169645.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400169645.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400169645.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400169645.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400169645.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400169645.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400169645.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400169645.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400169645.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400169645.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400169645.hg.1 | TC1400009422.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |

RAB15
Genomic Map