Assay Details
| Gene Symbol: LOC101929282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110244.1 | 3 - 4 | 425 | 78 | ||
| NR_110245.1 | 2 - 3 | 309 | 78 | |||
| NR_110246.1 | 2 - 3 | 350 | 78 | |||
| NR_110247.1 | 2 - 3 | 350 | 78 | |||
| GenBank mRNA | AA932755.1 | - | 2 - 3 | 78 | ||
| AA968441.1 | - | 2 - 3 | 78 | |||
| AI688708.1 | - | 2 - 3 | 78 | |||
| AW182822.1 | - | 1 - 2 | 78 | |||
| BF573640.1 | - | 2 - 3 | 251 | 78 | ||
| BX100556.1 | - | 2 - 3 | 180 | 78 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000423428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000423428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000423428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000423428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000423428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000423428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000423428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000443811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000443811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000443811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000443811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000443811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | ENST00000443811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009651.hg.1 | NR_110247.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000413247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000423428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000423428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000423428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000423428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000423428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000423428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000423428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000427615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000440668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000443811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000443811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000443811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000443811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000443811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | ENST00000443811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110247.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110247.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110247.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110247.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110247.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110247.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009651.hg.1 | NR_110247.1 | Refseq |

LOC101929282
Genomic Map