Assay Details
| Gene Symbol: LOC101927230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110806.1 | 2 - 3 | 138 | 72 | ||
| GenBank mRNA | AW104974.1 | - | 1 - 2 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927230| Gene Symbol: LOC101927230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110806.1 | 2 - 3 | 138 | 72 | ||
| GenBank mRNA | AW104974.1 | - | 1 - 2 | 72 | ||
Set Membership: |
|