Assay Details
Gene Symbol: FENDRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_036444.1 | 1 - 2 | 204 | 116 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16001325.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000593604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000593604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000593604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000593604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000593604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000593604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000593604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000594398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000594398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000594398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000594398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000594398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000594398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000595886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000595886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000595886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000595886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000595886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000595886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000595886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000595886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000595886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000595886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000595886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000595886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000595886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000597578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000597578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000597578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000597578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000597578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000597578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000598996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000598996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000598996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000598996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000598996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000598996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000598996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000598996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000598996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000598996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000598996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000598996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000599749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000599749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000599749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000599749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000599749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000599749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000599749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000599749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000599749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000599749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000599749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000599749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000599749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000599749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000599749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000599749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000600553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000600553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000600553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000600553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000600553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | ENST00000600553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_033925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_033925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_033925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_033925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_033925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_033925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_033925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_033925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_033925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_033925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_033925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_033925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_033925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_036444.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_036444.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_036444.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_036444.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_036444.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_036444.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_036444.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011143.hg.1 | NR_036444.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 230747 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 232975 | - | - |
