Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
ENST00000432035.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
ENST00000432035.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
ENST00000432035.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
ENST00000432035.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
ENST00000432035.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
ENST00000443990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
ENST00000443990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
ENST00000443990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
ENST00000545770.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
ENST00000545770.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
ENST00000545770.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
ENST00000545770.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
ENST00000545770.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
ENST00000545770.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
NM_001080528.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
NM_001080528.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
NM_001080528.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
NM_001080528.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
NM_001080528.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011055.hg.1 |
NM_001080528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
ENST00000432035.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
ENST00000432035.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
ENST00000432035.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
ENST00000432035.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
ENST00000432035.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
ENST00000443990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
ENST00000443990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
ENST00000443990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
ENST00000545770.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
ENST00000545770.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
ENST00000545770.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
ENST00000545770.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
ENST00000545770.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
ENST00000545770.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
NM_001080528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
NM_001080528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
NM_001080528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
NM_001080528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
NM_001080528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011055.hg.1 |
NM_001080528.2 |
Refseq |