Assay Details
| Gene Symbol: ZAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017004867.1 | XP_016860356.1 | 1 - 2 | 1290 | 91 | |
| XM_017004868.1 | XP_016860357.1 | 1 - 2 | 1290 | 91 | ||
| XM_017004869.1 | XP_016860358.1 | 1 - 2 | 1290 | 91 | ||
| XM_017004870.1 | XP_016860359.1 | 1 - 2 | 1290 | 91 | ||
More Information
Set Membership: |
|

ZAP70
Genomic Map