| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
ENST00000541604.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
ENST00000541604.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
ENST00000541604.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
ENST00000541604.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
ENST00000541604.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
ENST00000541604.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
ENST00000541604.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009200.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011890.hg.1 |
ENST00000372851.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011890.hg.1 |
ENST00000541604.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011890.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011890.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011890.hg.1 |
TC20000870.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
ENST00000541604.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
ENST00000541604.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
ENST00000541604.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
ENST00000541604.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
ENST00000541604.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
ENST00000541604.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
ENST00000541604.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009200.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164369.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164369.hg.1 |
ENST00000541604.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164369.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164369.hg.1 |
NM_182970.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164369.hg.1 |
TC2000009200.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000164369.hg.1 |
ENST00000372851.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164369.hg.1 |
ENST00000541604.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164369.hg.1 |
NM_001205317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164369.hg.1 |
NM_182970.3 |
Refseq |