Assay Details
Gene Symbol: C4BPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000715.3 | NP_000706.1 | 5 - 6 | 714 | 80 | |
XM_005273251.1 | XP_005273308.1 | 5 - 6 | 670 | 80 | ||
XM_005273252.4 | XP_005273309.1 | 5 - 6 | 846 | 80 | ||
GenBank mRNA | AK303793.1 | - | 6 - 7 | 525 | 80 | |
AK303798.1 | - | 5 - 6 | 664 | 80 | ||
AK313164.1 | - | 5 - 6 | 660 | 80 | ||
BC022312.1 | - | 5 - 6 | 714 | 4688428 | 80 | |
M31452.1 | - | 5 - 6 | 658 | 80 | ||
X02865.1 | - | 3 - 4 | 283 | 80 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001733.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01027089.hg.1 | TC01001733.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01027089.hg.1 | ENST00000367070.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01027089.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011403.hg.1 | ENST00000442684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011403.hg.1 | ENST00000442684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011403.hg.1 | ENST00000442684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011403.hg.1 | ENST00000442684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011403.hg.1 | ENST00000442684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011403.hg.1 | ENST00000442684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011403.hg.1 | ENST00000415474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011403.hg.1 | ENST00000442684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011403.hg.1 | ENST00000442684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011403.hg.1 | ENST00000442684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011403.hg.1 | ENST00000442684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011403.hg.1 | ENST00000442684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011403.hg.1 | ENST00000442684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100183884.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100183884.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100183884.hg.1 | ENST00000367070.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100183884.hg.1 | NM_000715.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100183884.hg.1 | TC0100011402.hg.1 | Clariom D TC |
