Assay Details
| Gene Symbol: CTSV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333.3 | NP_001324.2 | 1 - 2 | 237 | 144 | |
| GenBank mRNA | AY358641.1 | - | 1 - 2 | 237 | 144 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY358641.1, NM_001333.3 transcripts.
Set Membership: |
|

CTSV
Genomic Map