Assay Details
| Gene Symbol: DPYSL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006426.2 | NP_006417.2 | 9 - 10 | 1119 | 103 | |
| XM_005252658.3 | XP_005252715.1 | 9 - 10 | 1108 | 103 | ||
| XM_011539125.2 | XP_011537427.1 | 10 - 11 | 1122 | 103 | ||
| XM_017015482.1 | XP_016870971.1 | 9 - 10 | 2383 | 103 | ||
| XM_017015483.1 | XP_016870972.1 | 10 - 11 | 2536 | 103 | ||
| GenBank mRNA | AB006713.1 | - | 9 - 10 | 1113 | 103 | |
| BC029280.1 | - | 9 - 10 | 1003 | 103 | ||
| BC136329.1 | - | 9 - 10 | 988 | 9020316 | 103 | |
| BC136330.1 | - | 9 - 10 | 988 | 9020317 | 103 | |
| Y10976.1 | - | 9 - 10 | 996 | 103 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000941.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000471544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000471544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000471544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011550.hg.1 | ENST00000338492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011550.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011550.hg.1 | ENST00000368629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011550.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011550.hg.1 | TC10000941.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011551.hg.1 | ENST00000338492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011551.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011551.hg.1 | ENST00000368629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011551.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011551.hg.1 | TC10000941.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000471544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000471544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000471544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164210.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164210.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164210.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164210.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164210.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164210.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164210.hg.1 | TC1000009397.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000164211.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164211.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164211.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164211.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164211.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164211.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164211.hg.1 | TC1000009397.hg.1 | Clariom D TC |

DPYSL4
Genomic Map