Assay Details
| Gene Symbol: DHX9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U03643.1 | - | 2 - 3 | 265 | 144 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the U03643.1 transcripts.
Set Membership: |
|

DHX9
Genomic Map