Assay Details
| Gene Symbol: RBM46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277171.1 | NP_001264100.1 | 1 - 2 | 168 | 96 | |
| NM_144979.4 | NP_659416.1 | 1 - 2 | 168 | 96 | ||
| XM_006714119.2 | XP_006714182.1 | 1 - 2 | 165 | 96 | ||
| XM_006714120.1 | XP_006714183.1 | 1 - 2 | 165 | 96 | ||
| XM_011531697.2 | XP_011529999.1 | 1 - 2 | 165 | 96 | ||
| GenBank mRNA | AK302720.1 | - | 1 - 2 | 105 | 96 | |
| BC028588.2 | - | 1 - 2 | 92 | 4830259 | 96 | |
| DB088314.1 | - | 1 - 2 | 167 | 96 | ||
| DB449155.1 | - | 1 - 2 | 168 | 96 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000779.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK302720.1,BC028588.2,NM_001277171.1,NM_144979.4,XM_006714119.2,XM_006714120.1,XM_011531697.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04010767.hg.1 | TC04000779.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010767.hg.1 | ENST00000281722.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010767.hg.1 | ENST00000510397.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010767.hg.1 | ENST00000514866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010767.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000512640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000512640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000512640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009074.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010763.hg.1 | TC04000779.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010763.hg.1 | ENST00000281722.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010763.hg.1 | ENST00000514866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010763.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010764.hg.1 | TC04000779.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010764.hg.1 | ENST00000281722.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010764.hg.1 | ENST00000514866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010764.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162227.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162227.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162227.hg.1 | ENST00000512640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162227.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162227.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162227.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162227.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162227.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162227.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162227.hg.1 | ENST00000512640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162227.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162227.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162227.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162227.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162227.hg.1 | TC0400009074.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400162222.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162222.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162222.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162222.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162222.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162222.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162222.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162222.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162222.hg.1 | TC0400009074.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400162223.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162223.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162223.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162223.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162223.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162223.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162223.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162223.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162223.hg.1 | TC0400009074.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000281722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000510397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000512640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000512640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000512640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | ENST00000514866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_001277173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_144979.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009074.hg.1 | NM_144979.4 | Refseq |

RBM46
Genomic Map