Assay Details
| Gene Symbol: TBPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253676.1 | NP_001240605.1 | 3 - 4 | 821 | 77 | |
| NM_004865.3 | NP_004856.1 | 3 - 4 | 523 | 77 | ||
| XM_017011513.1 | XP_016867002.1 | 5 - 6 | 1096 | 77 | ||
| GenBank mRNA | AB020881.1 | - | 3 - 4 | 438 | 77 | |
| AF130312.1 | - | 3 - 4 | 466 | 77 | ||
| AF136570.1 | - | 3 - 4 | 470 | 77 | ||
| AK292320.1 | - | 3 - 4 | 776 | 77 | ||
| AK313645.1 | - | 3 - 4 | 491 | 77 | ||
| BC000381.2 | - | 4 - 5 | 491 | 2820491 | 77 | |
| BC017559.2 | - | 3 - 4 | 434 | 3911352 | 77 | |
| BI560657.1 | - | 3 - 4 | 528 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001002.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000367869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000367869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000367869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000367869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000367869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000367869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000367869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000416965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000416965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000416965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000416965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000416965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000416965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000416965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000416965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000457715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000457715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000457715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000457715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000457715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000457715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000477527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000477527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000477527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000477527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000481694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000481694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000481694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000481694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011836.hg.1 | TC06001002.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011836.hg.1 | ENST00000237264.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011836.hg.1 | ENST00000367871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011836.hg.1 | ENST00000477527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011836.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011836.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000367869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000367869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000367869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000367869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000367869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000367869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000367869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000416965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000416965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000416965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000416965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000416965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000416965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000416965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000416965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000457715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000457715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000457715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000457715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000457715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000457715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000477527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000477527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000477527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000477527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000481694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000481694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000481694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000481694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009508.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600165264.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165264.hg.1 | ENST00000367869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165264.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165264.hg.1 | ENST00000416965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165264.hg.1 | ENST00000477527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165264.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165264.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600165264.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600165264.hg.1 | ENST00000237264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165264.hg.1 | ENST00000367869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165264.hg.1 | ENST00000367871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165264.hg.1 | ENST00000416965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165264.hg.1 | ENST00000477527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165264.hg.1 | ENST00000613034.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600165264.hg.1 | NM_001253676.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600165264.hg.1 | NM_004865.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600165264.hg.1 | TC0600009508.hg.1 | Clariom D TC |

TBPL1
Genomic Map