Assay Details
| Gene Symbol: ZNF207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032293.2 | NP_001027464.1 | 6 - 7 | 754 | 61 | |
| NM_001098507.1 | NP_001091977.1 | 6 - 7 | 754 | 61 | ||
| XM_011525185.2 | XP_011523487.1 | 6 - 7 | 886 | 61 | ||
| XM_017025019.1 | XP_016880508.1 | 6 - 7 | 775 | 61 | ||
| GenBank mRNA | AB209215.1 | - | 6 - 7 | 712 | 61 | |
| AK226172.1 | - | 6 - 7 | 727 | 61 | ||
| AK298950.1 | - | 6 - 7 | 558 | 61 | ||
| AK309979.1 | - | 7 - 8 | 925 | 61 | ||
| AL834501.1 | - | 3 - 4 | 146 | 61 | ||
| BC002372.2 | - | 6 - 7 | 754 | 2822814 | 61 | |
| BC008023.1 | - | 6 - 7 | 420 | 3538197 | 61 | |
| BT007365.1 | - | 6 - 7 | 309 | 61 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ZNF207
Genomic Map