Assay Details
| Gene Symbol: SLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032214.3 | NP_115590.1 | 1 - 2 | 378 | 60 | |
| NM_175077.2 | NP_778252.1 | 1 - 2 | 378 | 60 | ||
| XM_017028098.1 | XP_016883587.1 | 1 - 2 | 369 | 60 | ||
| GenBank mRNA | AK291513.1 | - | 1 - 2 | 358 | 60 | |
| BC042041.1 | - | 1 - 2 | 317 | 4429896 | 60 | |
| BQ052468.1 | - | 1 - 2 | 43 | 60 | ||
| DA983348.1 | - | 1 - 2 | 378 | 60 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000815.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK291513.1,BC042041.1,NM_032214.3,NM_175077.2,XM_017028098.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
positive regulation of signal transduction
B cell mediated immunity
intracellular receptor signaling pathway
T cell activation
regulation of immune response
negative regulation of calcium-mediated signaling
antigen receptor-mediated signaling pathway
negative regulation of B cell activation
SH3/SH2 adaptor activity
protein binding
protein N-terminus binding
negative regulation of transcription from RNA polymerase II promoter
positive regulation of signal transduction
B cell mediated immunity
intracellular receptor signaling pathway
T cell activation
regulation of immune response
negative regulation of calcium-mediated signaling
antigen receptor-mediated signaling pathway
negative regulation of B cell activation
SH3/SH2 adaptor activity
protein binding
protein N-terminus binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20011169.hg.1 | ENST00000262866.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011169.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011169.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011169.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011169.hg.1 | TC20000815.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011171.hg.1 | ENST00000262866.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011171.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011171.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011171.hg.1 | TC20000815.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011168.hg.1 | ENST00000262866.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011168.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011168.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011168.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011168.hg.1 | TC20000815.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011170.hg.1 | ENST00000262866.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011170.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011170.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011170.hg.1 | TC20000815.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163084.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163084.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163084.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163084.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163084.hg.1 | TC2000009013.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000163084.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163084.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163084.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163084.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163085.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163085.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163085.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163085.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163085.hg.1 | TC2000009013.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000163085.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163085.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163085.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163085.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163086.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163086.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163086.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163086.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163086.hg.1 | TC2000009013.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000163086.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163086.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163086.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163086.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | ENST00000360672.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009013.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163087.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163087.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163087.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163087.hg.1 | TC2000009013.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000163087.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163087.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163087.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163088.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163088.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163088.hg.1 | NM_175077.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163088.hg.1 | TC2000009013.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000163088.hg.1 | ENST00000262866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163088.hg.1 | NM_032214.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163088.hg.1 | NM_175077.2 | Refseq |

SLA2
Genomic Map