Assay Details
| Gene Symbol: PAPPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020318.2 | NP_064714.2 | 2 - 3 | 2085 | 67 | |
| NM_021936.2 | NP_068755.2 | 2 - 3 | 2085 | 67 | ||
| XM_005245422.3 | XP_005245479.1 | 2 - 3 | 2132 | 67 | ||
| XM_011509857.1 | XP_011508159.1 | 3 - 4 | 2002 | 67 | ||
| XM_011509858.2 | XP_011508160.1 | 2 - 3 | 2345 | 67 | ||
| XM_017002023.1 | XP_016857512.1 | 3 - 4 | 2183 | 67 | ||
| XM_017002024.1 | XP_016857513.1 | 3 - 4 | 2230 | 67 | ||
| XM_017002025.1 | XP_016857514.1 | 2 - 3 | 2336 | 67 | ||
| GenBank mRNA | AF311940.1 | - | 1 - 2 | 921 | 67 | |
| AF342989.1 | - | 2 - 3 | 2085 | 67 | ||
| AF342990.1 | - | 2 - 3 | 2085 | 67 | ||
| AJ278348.1 | - | 1 - 2 | 884 | 67 | ||
| BC117193.1 | - | 1 - 2 | 1693 | 67 | ||
| BC117195.1 | - | 1 - 2 | 1694 | 67 | ||
| BC152552.1 | - | 2 - 3 | 2073 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001534.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01023659.hg.1 | TC01001534.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023659.hg.1 | ENST00000367661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023659.hg.1 | ENST00000367662.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023659.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023659.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000493665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000493665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023658.hg.1 | TC01001534.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023658.hg.1 | ENST00000367661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023658.hg.1 | ENST00000367662.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023658.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023658.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178692.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178692.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178692.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178692.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178692.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178692.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178692.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178692.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178692.hg.1 | TC0100010713.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100178694.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178694.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178694.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178694.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178694.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178694.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178694.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178694.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178694.hg.1 | TC0100010713.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000493665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000493665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 223045 | - | - |

PAPPA2
Genomic Map