| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001647.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001647.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001647.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001647.hg.1 |
TC20000127.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009896.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001645.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001645.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001645.hg.1 |
TC20000127.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001648.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001648.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001648.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001648.hg.1 |
TC20000127.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001649.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001649.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001649.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001649.hg.1 |
TC20000127.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2000169441.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169441.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169441.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000169441.hg.1 |
TC2000009896.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000169441.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169441.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169441.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009896.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000169442.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169442.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169442.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000169442.hg.1 |
TC2000009896.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000169442.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169442.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169442.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000169439.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169439.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000169439.hg.1 |
TC2000009896.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000169439.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169439.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000169443.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169443.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169443.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000169443.hg.1 |
TC2000009896.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000169443.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169443.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169443.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000169444.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169444.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169444.hg.1 |
NM_080820.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000169444.hg.1 |
TC2000009896.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000169444.hg.1 |
ENST00000377452.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169444.hg.1 |
ENST00000494921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000169444.hg.1 |
NM_080820.5 |
Refseq |