Assay Details
| Gene Symbol: ZMAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022470.3 | NP_071915.1 | 3 - 4 | 727 | 62 | |
| NM_152240.2 | NP_689426.1 | 4 - 5 | 848 | 62 | ||
| XM_005247706.2 | XP_005247763.1 | 3 - 4 | 1138 | 62 | ||
| XM_011513073.1 | XP_011511375.1 | 4 - 5 | 1259 | 62 | ||
| GenBank mRNA | AF355465.1 | - | 3 - 4 | 536 | 62 | |
| AK022358.1 | - | 3 - 4 | 654 | 62 | ||
| AK122768.1 | - | 4 - 5 | 737 | 62 | ||
| AY037945.1 | - | 2 - 3 | 426 | 62 | ||
| BC002896.2 | - | 3 - 4 | 655 | 3943641 | 62 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03002034.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03036040.hg.1 | TC03002034.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036040.hg.1 | ENST00000311417.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036040.hg.1 | ENST00000432729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036040.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036040.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000414084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000414084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000414084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000414084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000414084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000414084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036039.hg.1 | TC03002034.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036039.hg.1 | ENST00000311417.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036039.hg.1 | ENST00000432729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036039.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036039.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198767.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198767.hg.1 | ENST00000414084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198767.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198767.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198767.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198767.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198767.hg.1 | ENST00000414084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198767.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198767.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198767.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198767.hg.1 | TC0300013255.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000414084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000414084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000414084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000414084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000414084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000414084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013255.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198766.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198766.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198766.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198766.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198766.hg.1 | ENST00000311417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198766.hg.1 | ENST00000432729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198766.hg.1 | NM_022470.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198766.hg.1 | NM_152240.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198766.hg.1 | TC0300013255.hg.1 | Clariom D TC |

ZMAT3
Genomic Map