| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X016914.hg.1 |
TC0X001253.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X016914.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X016914.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010445.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010445.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010445.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010445.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010445.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010445.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010445.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010445.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010445.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010445.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010445.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010445.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010445.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010445.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X016916.hg.1 |
TC0X001253.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X016916.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X016916.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X016915.hg.1 |
TC0X001253.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X016915.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X016915.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171102.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171102.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171102.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171102.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171102.hg.1 |
TC0X00010445.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00010445.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010445.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010445.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010445.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010445.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010445.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010445.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010445.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010445.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010445.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010445.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010445.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010445.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010445.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171101.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171101.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171101.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171101.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171101.hg.1 |
TC0X00010445.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00171103.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171103.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171103.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171103.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171103.hg.1 |
TC0X00010445.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00171104.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171104.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171104.hg.1 |
ENST00000372588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171104.hg.1 |
NM_153448.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171104.hg.1 |
TC0X00010445.hg.1 |
Clariom D TC |