Assay Details
Gene Symbol: URB1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026845.1 | 1 | 241 | 86 | ||
GenBank mRNA | AK026376.1 | - | 1 | 219 | 86 | |
BC007928.1 | - | 1 | 247 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC21000116.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11354583 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32014194 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32014187 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11354575 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32014192 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11354580 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11354589 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11354587 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32014190 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11354588 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32014196 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17231178 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17231177 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11354576 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 27328 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 48028 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P29029 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 12389 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 13204 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 4458 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P29029 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P29029 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354588 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17231175 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014187 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014197 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014205 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17231177 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354575 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014203 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354589 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17231178 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354590 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014201 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014193 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014190 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354581 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014192 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354583 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17231176 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354584 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014191 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354587 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354577 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014194 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354576 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014196 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354580 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32014187 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32014194 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32014192 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32014190 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11354588 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32014196 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11354575 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11354576 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11354589 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17231177 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354584 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014196 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014193 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014205 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17231178 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354590 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354587 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17231176 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354575 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014201 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17231175 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014187 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354589 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014203 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014194 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014191 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014192 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354580 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17231177 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354577 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354581 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014197 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354576 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354583 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11354588 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32014190 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11354589 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11354575 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11354588 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32014196 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32014187 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32014194 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11354576 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32014192 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32014190 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17231177 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P29023 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P29029 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P110471 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 43755 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P110471 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11354575 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11354577 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11354585 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11354576 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11354579 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11354581 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11354594 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11354588 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11354546 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11354591 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11354589 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11354590 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17231178 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11354574 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P110471 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8068201 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8068200 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 227424_PM_x_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR21000737.hg.1 | ENST00000534991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21000737.hg.1 | NR_026845.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21000737.hg.1 | TC21000116.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2100006920.hg.1 | ENST00000534991.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100006920.hg.1 | NR_026845.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100146473.hg.1 | ENST00000534991.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100146473.hg.1 | NR_026845.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100146473.hg.1 | TC2100006920.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100146473.hg.1 | ENST00000534991.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100146473.hg.1 | NR_026845.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100006920.hg.1 | ENST00000534991.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100006920.hg.1 | NR_026845.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 227424_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 227424_x_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227424_x_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227424_x_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227424_x_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227424_x_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227424_x_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227424_x_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227424_x_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227424_x_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227424_x_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227424_x_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227424_x_at10 | - | - |
Human Genome U95 Set | 53179_PM_at | - | - |
Human Genome U95 Set | 53179_at | - | - |
Human Genome U133 Set | 227424_x_at | - | - |
Human Genome U133 Set | 227424_PM_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019159 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019159 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 147385 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1728403 | NR_026845.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1728403 | AI369236.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1728403 | AK026376.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1728403 | BC007928.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1728403 | NR_026845.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1728403 | AI369236.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1728403 | AK026376.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1728403 | BC007928.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1728403 | NR_026845.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1728403 | AI369236.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1728403 | AK026376.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1728403 | BC007928.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1728403 | NR_026845.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1728403 | AI369236.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1728403 | AK026376.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1728403 | BC007928.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1728403 | NR_026845.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1728403 | AI369236.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1728403 | AK026376.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1728403 | BC007928.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1728403 | NR_026845.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1728403 | AI369236.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1728403 | AK026376.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1728403 | BC007928.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1728403 | NR_026845.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1728403 | AI369236.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1728403 | AK026376.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1728403 | BC007928.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1728403 | NR_026845.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1728403 | AI369236.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1728403 | AK026376.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1728403 | BC007928.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1728403 | NR_026845.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1728403 | AI369236.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1728403 | AK026376.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1728403 | BC007928.1 | human_transcriptome |
