Assay Details
| Gene Symbol: SHANK2-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073536.1 | 1 - 2 | 81 | 68 | ||
| GenBank mRNA | BC004224.2 | - | 1 - 2 | 81 | 3537945 | 68 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC004224.2 transcripts.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28280638 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864390 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864332 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864364 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864373 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28280584 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28280606 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16469869 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28280596 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864396 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28280610 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864376 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864359 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864342 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864331 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864379 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28280627 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28280599 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864381 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864346 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864329 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864392 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864398 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28280639 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864370 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864380 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864344 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864351 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864348 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864340 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16469868 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864353 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16469875 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864386 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16469876 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28280646 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864341 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28280594 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864395 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864371 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864330 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864374 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864368 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864334 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864363 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28280633 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864335 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28280630 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28280632 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16469873 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28280585 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864383 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07864357 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28280583 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 27265 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 59997 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3397161 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P244447 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7942230 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7942231 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7942229 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7942228 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 224139_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008289.hg.1 | ENST00000307548.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008289.hg.1 | ENST00000307548.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008289.hg.1 | ENST00000307548.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008289.hg.1 | NR_073536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008289.hg.1 | NR_073536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008289.hg.1 | NR_073536.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161148.hg.1 | ENST00000307548.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100161148.hg.1 | NR_073536.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161148.hg.1 | ENST00000307548.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100161148.hg.1 | NR_073536.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161148.hg.1 | TC1100008289.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100008289.hg.1 | ENST00000307548.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008289.hg.1 | ENST00000307548.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008289.hg.1 | ENST00000307548.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008289.hg.1 | NR_073536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008289.hg.1 | NR_073536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008289.hg.1 | NR_073536.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161149.hg.1 | ENST00000307548.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100161149.hg.1 | NR_073536.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161149.hg.1 | ENST00000307548.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100161149.hg.1 | NR_073536.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161149.hg.1 | TC1100008289.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100161150.hg.1 | ENST00000307548.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100161150.hg.1 | NR_073536.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161150.hg.1 | ENST00000307548.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100161150.hg.1 | NR_073536.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161150.hg.1 | TC1100008289.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 224139_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 224139_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224139_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224139_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224139_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224139_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224139_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224139_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224139_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224139_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224139_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224139_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224139_at6 | - | - |
| Human Genome U133 Set | 224139_at | - | - |
| Human Genome U133 Set | 224139_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 206102 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1669579 | BC004224.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2168079 | BC004224.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1669579 | BC004224.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1669579 | BC004224.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2168079 | BC004224.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1669579 | BC004224.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2168079 | BC004224.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2168079 | BC004224.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1669579 | BC004224.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2168079 | BC004224.2 | human_transcriptome |

SHANK2-AS3
Genomic Map