| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015626.hg.1 |
TC09001218.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015626.hg.1 |
ENST00000376834.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015626.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000376830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000376830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000376830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000376830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000376830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000451153.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000451153.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000451153.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000451153.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000451153.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000455336.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000455336.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
ENST00000615099.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171448.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171448.hg.1 |
ENST00000451153.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171448.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171448.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171448.hg.1 |
ENST00000451153.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171448.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171448.hg.1 |
TC0900010444.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000376830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000376830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000376830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000376830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000376830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000376834.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000451153.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000451153.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000451153.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000451153.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000451153.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000455336.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000455336.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
ENST00000615099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010444.hg.1 |
NM_152420.2 |
Refseq |