Assay Details
| Gene Symbol: C6orf89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286635.1 | NP_001273564.1 | 6 - 7 | 985 | 89 | |
| NM_001286636.1 | NP_001273565.1 | 7 - 8 | 1049 | 89 | ||
| NM_001286637.1 | NP_001273566.1 | 6 - 7 | 926 | 89 | ||
| NM_152734.3 | NP_689947.2 | 5 - 6 | 885 | 89 | ||
| XM_011514377.2 | XP_011512679.1 | 8 - 9 | 1748 | 89 | ||
| XM_017010432.1 | XP_016865921.1 | 8 - 9 | 1202 | 89 | ||
| XM_017010433.1 | XP_016865922.1 | 8 - 9 | 1203 | 89 | ||
| XM_017010434.1 | XP_016865923.1 | 7 - 8 | 1441 | 89 | ||
| GenBank mRNA | AJ420498.1 | - | 5 - 6 | 892 | 88 | |
| AK058086.1 | - | 7 - 8 | 1049 | 89 | ||
| AK172780.1 | - | 5 - 6 | 794 | 89 | ||
| AK300351.1 | - | 6 - 7 | 863 | 89 | ||
| AK315984.1 | - | 6 - 7 | 835 | 89 | ||
| AY358637.1 | - | 6 - 7 | 859 | 89 | ||
| BC009380.2 | - | 1 - 2 | 99 | 89 | ||
| BC023605.2 | - | 5 - 6 | 743 | 89 | ||
| BC095423.1 | - | 6 - 7 | 898 | 30348810 | 89 | |
| BC105946.1 | - | 5 - 6 | 792 | 40032607 | 89 | |
| BC105947.1 | - | 5 - 6 | 793 | 40032604 | 89 | |
| BC107489.1 | - | 5 - 6 | 793 | 40032608 | 89 | |
| BX538108.1 | - | 5 - 6 | 778 | 89 | ||
| BX647220.1 | - | 6 - 7 | 857 | 89 | ||
| FR832871.1 | - | 6 - 7 | 929 | 89 | ||
| FR832872.1 | - | 5 - 6 | 914 | 89 | ||
More Information
Set Membership: |
|

C6orf89
Genomic Map