Assay Details
| Gene Symbol: C6orf89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286635.1 | NP_001273564.1 | 7 - 8 | 1106 | 70 | |
| NM_001286636.1 | NP_001273565.1 | 8 - 9 | 1170 | 70 | ||
| NM_001286637.1 | NP_001273566.1 | 7 - 8 | 1047 | 70 | ||
| NM_152734.3 | NP_689947.2 | 6 - 7 | 1006 | 70 | ||
| XM_011514377.2 | XP_011512679.1 | 9 - 10 | 1869 | 70 | ||
| XM_017010432.1 | XP_016865921.1 | 9 - 10 | 1323 | 70 | ||
| XM_017010433.1 | XP_016865922.1 | 9 - 10 | 1324 | 70 | ||
| XM_017010434.1 | XP_016865923.1 | 8 - 9 | 1562 | 70 | ||
| GenBank mRNA | AJ420498.1 | - | 6 - 7 | 1012 | 70 | |
| AK058086.1 | - | 8 - 9 | 1170 | 70 | ||
| AK172780.1 | - | 6 - 7 | 915 | 70 | ||
| AK300351.1 | - | 7 - 8 | 984 | 70 | ||
| AK315984.1 | - | 7 - 8 | 956 | 70 | ||
| AY358637.1 | - | 7 - 8 | 980 | 70 | ||
| BC009380.2 | - | 2 - 3 | 220 | 70 | ||
| BC023605.2 | - | 6 - 7 | 864 | 70 | ||
| BC095423.1 | - | 7 - 8 | 1019 | 30348810 | 70 | |
| BC105946.1 | - | 6 - 7 | 913 | 40032607 | 70 | |
| BC105947.1 | - | 6 - 7 | 914 | 40032604 | 70 | |
| BC107489.1 | - | 6 - 7 | 914 | 40032608 | 70 | |
| BX538108.1 | - | 6 - 7 | 899 | 70 | ||
| BX647220.1 | - | 7 - 8 | 978 | 70 | ||
| FR832871.1 | - | 7 - 8 | 1050 | 70 | ||
| FR832872.1 | - | 6 - 7 | 1035 | 70 | ||
More Information
Set Membership: |
|

C6orf89
Genomic Map