Assay Details
| Gene Symbol: SP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173467.2 | NP_001166938.1 | 2 - 3 | 311 | 135 | |
| NM_152860.1 | NP_690599.1 | 1 - 2 | 133 | 135 | ||
| GenBank mRNA | AF466179.1 | - | 1 - 2 | 134 | 135 | |
| AF477981.1 | - | 1 - 2 | 133 | 135 | ||
| AK128520.1 | - | 2 - 3 | 310 | 135 | ||
| AY150673.1 | - | 1 - 2 | 72 | 135 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001548.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF466179.1,AF477981.1,AK128520.1,AY150673.1,NM_001173467.2,NM_152860.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
hematopoietic stem cell differentiation
positive regulation of stem cell differentiation
DNA binding
DEAD/H-box RNA helicase binding
metal ion binding
osteoblast differentiation
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
hematopoietic stem cell differentiation
positive regulation of stem cell differentiation
DNA binding
DEAD/H-box RNA helicase binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12020455.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020455.hg.1 | ENST00000536324.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020455.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020455.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020455.hg.1 | TC12001548.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020457.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020457.hg.1 | ENST00000536324.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020457.hg.1 | ENST00000537210.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020457.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020457.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020457.hg.1 | TC12001548.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020462.hg.1 | ENST00000536324.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020462.hg.1 | ENST00000537210.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020462.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020462.hg.1 | TC12001548.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000536324.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000536324.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000536324.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000536324.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000536324.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000536324.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000536324.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000536324.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000537210.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000537210.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000537210.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000537210.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000547755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000547755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | ENST00000547755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_001300837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_001300837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_001300837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_001300837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_001300837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_001300837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_001300837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_001300837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010799.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020458.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020458.hg.1 | ENST00000536324.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020458.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020458.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020458.hg.1 | TC12001548.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020460.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020460.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020460.hg.1 | TC12001548.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020456.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020456.hg.1 | ENST00000536324.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020456.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020456.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020456.hg.1 | TC12001548.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200182678.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200182678.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200182678.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200182678.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200182678.hg.1 | TC1200010799.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200182676.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200182676.hg.1 | ENST00000536324.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200182676.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200182676.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200182676.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200182676.hg.1 | ENST00000536324.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200182676.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200182676.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200182676.hg.1 | TC1200010799.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000303846.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000536324.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000536324.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000536324.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000536324.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000536324.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000536324.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000536324.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000536324.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000537210.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000537210.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000537210.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000537210.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000547755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000547755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | ENST00000547755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_001173467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_001300837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_001300837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_001300837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_001300837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_001300837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_001300837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_001300837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_001300837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_152860.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010799.hg.1 | NM_152860.1 | Refseq |

SP7
Genomic Map