Assay Details
| Gene Symbol: LOC107987235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017023960.1 | XP_016879449.1 | 1 | 848 | 106 | |
| Gene Symbol: LOC162137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK312764.1 | - | 1 | 720 | 106 | |
| BC029861.1 | - | 1 | 854 | 5167909 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988287 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526722 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988258 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526665 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526752 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988257 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526757 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988283 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988250 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526719 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526706 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988322 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526675 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988276 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988284 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988330 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526718 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526682 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988282 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988329 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526725 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988289 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526689 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988331 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526743 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526693 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526762 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526775 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526721 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988343 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526690 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526735 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526692 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988315 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988317 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526694 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526776 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988300 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526687 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526680 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988279 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526770 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988342 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526766 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526720 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526730 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988254 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988280 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988251 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526729 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988285 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526672 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988275 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16890456 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16890457 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988277 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526688 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988318 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526774 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988295 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988296 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988286 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526746 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988348 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526765 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988334 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526685 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526691 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526734 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988260 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526737 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988316 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526701 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526771 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988301 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526686 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988281 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526748 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526736 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526732 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526728 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988273 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526767 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526764 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988336 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526724 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526731 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988270 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988337 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526754 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988328 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526733 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P219418 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 35518 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P119381 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P119381 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 1634 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3373243 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P219418 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P119381 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P119381 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 99546 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 104615 | AK092883.1 | genbank_mrna_human |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 104615 | AK122584.1 | genbank_mrna_human |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 104615 | AK125635.1 | genbank_mrna_human |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 104615 | AX747788.1 | genbank_mrna_human |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 104615 | BC029917.1 | genbank_mrna_human |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 104615 | BX648550.1 | genbank_mrna_human |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 104615 | NM_152309.2 | refseq_mrna_human_NM |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P119381 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P855532 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1553914_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007655.hg.1 | ENST00000319817.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007655.hg.1 | ENST00000319817.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007655.hg.1 | ENST00000319817.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007655.hg.1 | ENST00000319817.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007655.hg.1 | ENST00000570988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010126.hg.1 | ENST00000567803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010126.hg.1 | ENST00000567803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007655.hg.1 | ENST00000319817.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007655.hg.1 | ENST00000319817.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007655.hg.1 | ENST00000319817.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007655.hg.1 | ENST00000319817.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007655.hg.1 | ENST00000570988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178149.hg.1 | TC1600010124.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010126.hg.1 | ENST00000567803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010126.hg.1 | ENST00000567803.1 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 1553914_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1553914_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553914_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553914_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553914_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553914_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553914_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553914_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553914_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553914_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553914_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553914_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553914_at11 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 225767 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1658111 | BC029861.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1658111 | BC029861.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1658111 | BC029861.1 | human_transcriptome |

LOC107987235
Genomic Map